missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxycarbonyl-2-nitrophenylboronic Acid (contains varying amounts of Anhydride) 98.0+%, TCI America™

Catalog No. E12031G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E12031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E12031G Supplier TCI America Supplier No. E12031G

Chemical Identifiers

CAS 5785-70-6
Molecular Formula C9H10BNO6
Molecular Weight (g/mol) 238.99
MDL Number MFCD02179460
InChI Key GCDAYMSNTGTFDC-UHFFFAOYSA-N
Synonym 4-ethoxycarbonyl-2-nitrophenyl boronic acid, ethyl 4-borono-3-nitrabenzoate, ethyl 4-borono-3-nitrobenzoate, 4-ethoxycarbonyl-2-nitrophenylboronic acid, 4-ethoxycarbonyl-2-nitrobenzeneboronic acid, 4-ethoxycarbonyl-2-nitrophenylboronicacid, acmc-1alqp, chembl91674, benzoic acid,4-borono-3-nitro-, 1-ethyl ester
PubChem CID 2773403
IUPAC Name (4-ethoxycarbonyl-2-nitrophenyl)boronic acid
SMILES B(C1=C(C=C(C=C1)C(=O)OCC)[N+](=O)[O-])(O)O

Specifications

Melting Point 125°C
Color White-Yellow
Quantity 1 g
Formula Weight 238.99
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Ethoxycarbonyl-2-nitrophenylboronic Acid (contains varying amounts of Anhydride)