missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxybenzoic Acid 98.0+%, TCI America™

Catalog No. E004525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E004525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E004525G Supplier TCI America Supplier No. E004525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 619-86-3
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00002545
InChI Key SHSGDXCJYVZFTP-UHFFFAOYSA-N
Synonym p-ethoxybenzoic acid, benzoic acid, 4-ethoxy, benzoic acid, p-ethoxy, 4-ethoxy-benzoic acid, unii-tzr23xeq7w, tzr23xeq7w, 4-ethoxybenzoicacid, 4ethoxybenzoic acid, pubchem2686, para-ethoxybenzoic acid
PubChem CID 12093
IUPAC Name 4-ethoxybenzoic acid
SMILES CCOC1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 200°C
Color White-Yellow
Quantity 25 g
Formula Weight 166.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Ethoxybenzoic Acid