missing translation for 'onlineSavingsMsg'
Learn More

4-(Dodecyloxy)benzoic Acid 98.0+%, TCI America™

Catalog No. D28595G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28595G 5 g
D285925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28595G Supplier TCI America Supplier No. D28595G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2312-15-4
Molecular Formula C19H30O3
Molecular Weight (g/mol) 306.45
MDL Number MFCD00002543
InChI Key ALQLYJHDBAKLBB-UHFFFAOYSA-N
Synonym 4-dodecyloxy benzoic acid, p-dodecyloxybenzoic acid, 4-n-dodecyloxybenzoic acid, 4-dodecyloxybenzoic acid, benzoic acid, 4-dodecyloxy, benzoic acid, p-dodecyloxy, p-dodecyloxy benzoic acid, 4-dodecyloxy benzoicacid, pubchem2685, acmc-1crse
PubChem CID 75330
IUPAC Name 4-(dodecyloxy)benzoic acid
SMILES CCCCCCCCCCCCOC1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 137°C
Color White
Quantity 5 g
Formula Weight 306.45
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Dodecyloxy)benzoic Acid