missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminopyridine, Reagent (DMAP), Reagents

Catalog No. 01337284
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337284 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01337284 Supplier Labchem Inc Supplier No. C2196050100C2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

4-Dimethylaminopyridine, Reagent (DMAP), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

CAS 1122-58-3
Boiling Range 162°C
Color White to Beige
Boiling Point 162°C
Packaging Glass Bottle
Molecular Formula C7H11N2
Quantity 100 g
Specific Gravity 1.012
Synonym DMAP, N,N-Dimethylpyridin-4-amine
InChI Key VHYFNPMBLIVWCW-UHFFFAOYSA-O
SMILES CN(C)C1=CC=[NH+]C=C1
IUPAC Name 4-(dimethylamino)pyridin-1-ium
Molecular Weight (g/mol) 123.18
Percent Purity 99%
Grade Reagent
Physical Form Solid
Chemical Name or Material 4-Dimethylaminopyridine, Reagent (DMAP)
Show More Show Less