missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminophenyl isothiocyanate, 97%

Catalog No. p-7044990
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0901104 2 g
AAL0901109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0901104 Supplier Thermo Scientific Chemicals Supplier No. L0901104

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2131-64-8 | C9H10N2S | 178.253 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2131-64-8
Molecular Formula C9H10N2S
Molecular Weight (g/mol) 178.253
MDL Number MFCD00041093
InChI Key HRDJPEMAGYHSJR-UHFFFAOYSA-N
PubChem CID 75047
IUPAC Name 4-isothiocyanato-N,N-dimethylaniline
SMILES CN(C)C1=CC=C(C=C1)N=C=S

Specifications

Melting Point 64°C to 70°C
Boiling Point 152°C (6 mmHg)
Odor Irritating
Quantity 2 g
UN Number UN2928
Beilstein 388270
Sensitivity Moisture sensitive
Synonym 4-dimethylamino phenyl isothiocyanate, 4-dimethylaminophenyl isothiocyanate, 4-n,n-dimethylamino phenyl isothiocyanate, p-dimethylamino phenyl isothiocyanate, benzenamine, 4-isothiocyanato-n,n-dimethyl, isothiocyanic acid, p-dimethylaminophenyl ester, p-n-dimethylamino phenyl isothiocyanate, 4-dimethylamino phenylisothiocyanate, benzenamine,4-isothiocyanato-n,n-dimethyl, isothiocyanic acid, p-dimethylamino phenyl ester
Formula Weight 178.25
Percent Purity 97%
Chemical Name or Material 4-Dimethylaminophenyl isothiocyanate
Show More Show Less
Hazard Category H301+H311-H314-H330-H335
Hazard Statement GHS H Statement
H300-H310-H330-H314-H318
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, CORROSIVE, ORGANIC, N.O.S.
EINECSNumber 218-360-4
RTECSNumber NX8500000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only