missing translation for 'onlineSavingsMsg'
Learn More

(4-Dimethylaminophenyl)di-tert-butylphosphine, TCI America™

Catalog No. D45311G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D45311G Supplier TCI America Supplier No. D45311G
Only null left

Specifications

Chemical Name or Material (4-Dimethylaminophenyl)di-tert-butylphosphine
CAS 932710-63-9
Melting Point 69°C
Color White-Yellow
Molecular Formula C16H28NP
Quantity 1 g
MDL Number MFCD09265102
Synonym 4-(Di-tert-butylphosphino)-N,N-dimethylaniline, APhos, A-taPhos, Amphos
InChI Key IQTHEAQKKVAXGV-UHFFFAOYSA-N
SMILES CC(C)(C)P(C1=CC=C(C=C1)N(C)C)C(C)(C)C
IUPAC Name 4-ditert-butylphosphanyl-N,N-dimethylaniline
Molecular Weight (g/mol) 265.381
PubChem CID 11714598
Formula Weight 265.38
Physical Form Crystalline Powder
Show More Show Less