missing translation for 'onlineSavingsMsg'
Learn More

4-(Dimethylamino)benzophenone 98.0+%, TCI America™

Catalog No. D25615G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25615G 5 g
D256125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D25615G Supplier TCI America Supplier No. D25615G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 530-44-9
Molecular Formula C15H15NO
Molecular Weight (g/mol) 225.291
MDL Number MFCD00008311
InChI Key BEUGBYXJXMVRFO-UHFFFAOYSA-N
Synonym 4-dimethylamino benzophenone, 4-dimethylaminobenzophenone, p-dimethylaminobenzophenone, p-benzoyl-n,n-dimethylaniline, 4-n,n-dimethylaminobenzophenone, p-dimethylamino benzophenone, 4-dimethylamino phenyl phenyl methanone, 4-benzoyl-n,n-dimethylaniline, methanone, 4-dimethylamino phenyl phenyl, benzophenone, 4-dimethylamino
PubChem CID 10737
IUPAC Name [4-(dimethylamino)phenyl]-phenylmethanone
SMILES CN(C)C1=CC=C(C=C1)C(=O)C2=CC=CC=C2

Specifications

Melting Point 90°C
Boiling Point 250°C
Quantity 5 g
Formula Weight 225.29
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Dimethylamino)benzophenone