missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylamino-2-methoxybenzaldehyde, 98%

Catalog No. AAB2359806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2359806 5 g
AAB2359803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2359806 Supplier Thermo Scientific Chemicals Supplier No. B2359806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 84562-48-1 | C10H13NO2 | 179.219 g/mol

4-Dimethylamino-2-methoxybenzaldehyde is used to produce 2,6-Dichloro-N'-(4-dimethylamino-2-methoxy-benzylidene)-benzene-1,4-diamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Dimethylamino-2-methoxybenzaldehyde is used to produce 2,6-Dichloro-N′-(4-dimethylamino-2-methoxy-benzylidene)-benzene-1,4-diamine.

Solubility
Slightly soluble 0.32 g/L at 25°C.

Notes
Air sensitive. Store in cool, dry conditions in well sealed containers. Store away from oxidizing agents.

Chemical Identifiers

CAS 84562-48-1
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.219
MDL Number MFCD00151814
InChI Key HGDRXADJVGVGBC-UHFFFAOYSA-N
PubChem CID 291350
IUPAC Name 4-(dimethylamino)-2-methoxybenzaldehyde
SMILES CN(C)C1=CC(=C(C=C1)C=O)OC

Specifications

Melting Point 58°C to 60°C
Quantity 5 g
Beilstein 2719181
Sensitivity Air Sensitive
Synonym 4-dimethylamino-2-methoxybenzaldehyde, 2-methoxy-4-dimethylaminobenzaldehyde, 4-dimethylamino-2-methoxy-benzaldehyde, benzaldehyde, 4-dimethylamino-2-methoxy, 2-methoxy-4-dimethylamino benzaldehyde, benzaldehyde,4-dimethylamino-2-methoxy, 2-methoxy-4-n,n-dimethylamino benzaldehyde
Solubility Information Slightly soluble 0.32g/L at 25°C.
Formula Weight 179.22
Percent Purity 98%
Chemical Name or Material 4-Dimethylamino-2-methoxybenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only