missing translation for 'onlineSavingsMsg'
Learn More

4'-(Difluoromethoxy)acetophenone 98.0+%, TCI America™

Catalog No. D47605G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47605G 5 g
D47601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47605G Supplier TCI America Supplier No. D47605G
Only null left

Chemical Identifiers

CAS 83882-67-1
Molecular Formula C9H8F2O2
Molecular Weight (g/mol) 186.158
MDL Number MFCD00042250
InChI Key GIGWRVLNOYPOIT-UHFFFAOYSA-N
PubChem CID 737356
IUPAC Name 1-[4-(difluoromethoxy)phenyl]ethanone
SMILES CC(=O)C1=CC=C(C=C1)OC(F)F

Specifications

Color Yellow
Quantity 5 g
Formula Weight 186.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4′-(Difluoromethoxy)acetophenone