missing translation for 'onlineSavingsMsg'
Learn More

4-(Diethylcarbamoyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. D46811G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D46811G Supplier TCI America Supplier No. D46811G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 4-(Diethylcarbamoyl)phenylboronic Acid (contains varying amounts of Anhydride)
CAS 389621-80-1
Melting Point 142°C
Color White
Molecular Formula C11H16BNO3
Quantity 1 g
MDL Number MFCD03411949
Synonym 4-n,n-diethylaminocarbonyl phenylboronic acid, 4-diethylcarbamoyl phenylboronic acid, 4-diethylcarbamoyl phenyl boronic acid, 4-diethylcarbamoyl benzeneboronic acid, boronic acid, 4-diethylamino carbonyl phenyl, 4-diethylaminocarbonyl phenylboronic acid, acmc-209j2q, amtb302, n,n-diethyl 4-boronobenzamide, 4-diethylcarbamoyl benzeneboronic acid
InChI Key ZCGVBHIMRVYWOH-UHFFFAOYSA-N
SMILES CCN(CC)C(=O)C1=CC=C(C=C1)B(O)O
IUPAC Name [4-(diethylcarbamoyl)phenyl]boronic acid
Molecular Weight (g/mol) 221.06
PubChem CID 2773375
Formula Weight 221.06
Physical Form Crystalline Powder
Show More Show Less