missing translation for 'onlineSavingsMsg'
Learn More

4-(Diethylcarbamoyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals

Catalog No. AAH5298606
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5298606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5298606 Supplier Thermo Scientific Chemicals Supplier No. H5298606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 389621-80-1 | Molecular Formula: C11H16BNO3 | Molecular Weight: 221.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 389621-80-1
Molecular Formula C11H16BNO3
Molecular Weight (g/mol) 221.06
MDL Number MFCD03411949
InChI Key ZCGVBHIMRVYWOH-UHFFFAOYSA-N
Synonym 4-n,n-diethylaminocarbonyl phenylboronic acid, 4-diethylcarbamoyl phenylboronic acid, 4-diethylcarbamoyl phenyl boronic acid, 4-diethylcarbamoyl benzeneboronic acid, boronic acid, 4-diethylamino carbonyl phenyl, 4-diethylaminocarbonyl phenylboronic acid, acmc-209j2q, amtb302, n,n-diethyl 4-boronobenzamide, 4-diethylcarbamoyl benzeneboronic acid
PubChem CID 2773375
IUPAC Name [4-(diethylcarbamoyl)phenyl]boronic acid
SMILES CCN(CC)C(=O)C1=CC=C(C=C1)B(O)O

Specifications

Melting Point 138°C to 142°C
Quantity 5 g
Formula Weight 221.06
Percent Purity 98%
Chemical Name or Material 4-(Diethylcarbamoyl)benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only