missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 4-(Dicyanomethylene)-2-methyl-6-(p-dimethylaminostyryl)-4H-pyran, 96+%

Catalog No. AC407072500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC407072500 250 mg
AC407070010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC407072500 Supplier Thermo Scientific Chemicals Supplier No. 407072500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 51325-91-8 | C19H17N3O | 303.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51325-91-8
Molecular Formula C19H17N3O
Molecular Weight (g/mol) 303.35
MDL Number MFCD00051341
InChI Key YLYPIBBGWLKELC-RMKNXTFCSA-N
Synonym unii-9s1wxc1zgn, dcm dye, dcm1, 9s1wxc1zgn, 4-dicyanomethylene-2-methyl-6-4-dimethylaminostyryl-4h-pyran, 4-dicyanomethylene-2-methyl-6-4-dimethylamino styryl-4h-pyran, e-2-2-4-dimethylamino styryl-6-methyl-4h-pyran-4-ylidene malononitrile, 2-2-4-dimethylamino phenyl ethenyl-6-methyl-4h-pyran-4-ylidene propanedinitrile, 2-2-4-dimethylamino styryl-6-methyl-4h-pyran-4-ylidene malononitrile
PubChem CID 688222
IUPAC Name 2-[2-[(E)-2-[4-(dimethylamino)phenyl]ethenyl]-6-methylpyran-4-ylidene]propanedinitrile
SMILES CC1=CC(=C(C#N)C#N)C=C(O1)C=CC2=CC=C(C=C2)N(C)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 212.0°C
Color Red
Flash Point 43°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 250 mg
Merck Index 15, 2841
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in benzene,toluene,chloroform
Formula Weight 303.35
Percent Purity 96+%
Grade Extra Pure
Physical Form Crystalline Needles or Powder
Chemical Name or Material 4-(Dicyanomethylene)-2-methyl-6-(p-dimethylaminostyryl)-4H-pyran, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Flammable solid.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Call a POISON CENTER or
EINECSNumber 257-137-6
RTECSNumber OO3746100
TSCA TSCA

RUO – Research Use Only