missing translation for 'onlineSavingsMsg'
Learn More

4-Diazo-N-benzyl-N-ethylaniline Chloride Zinc Chloride 98.0+%, TCI America™

Catalog No. D013825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D013825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D013825G Supplier TCI America Supplier No. D013825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15280-31-6
Molecular Formula C30H32Cl4N6Zn
Molecular Weight (g/mol) 683.81
MDL Number MFCD00070598
InChI Key GZWAXOPGCCVFJG-UHFFFAOYSA-J
Synonym N-Benzyl-N-ethyl-1,4-phenylenediamine Diazonium Chloride Zinc Chloride
PubChem CID 73555550
IUPAC Name zinc(2+) bis(4-[benzyl(ethyl)amino]benzene-1-diazonium) tetrachloride
SMILES [Cl-].[Cl-].[Cl-].[Cl-].[Zn++].CCN(CC1=CC=CC=C1)C1=CC=C(C=C1)[N+]#N.CCN(CC1=CC=CC=C1)C1=CC=C(C=C1)[N+]#N

Specifications

Quantity 25 g
Formula Weight 683.81
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Diazo-N-benzyl-N-ethylaniline Chloride Zinc Chloride