missing translation for 'onlineSavingsMsg'
Learn More

4-Cyclohexylphenol 98.0+%, TCI America™

Catalog No. C073325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C073325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C073325G Supplier TCI America Supplier No. C073325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1131-60-8
Molecular Formula C12H16O
Molecular Weight (g/mol) 176.259
MDL Number MFCD00035699
InChI Key OAHMVZYHIJQTQC-UHFFFAOYSA-N
Synonym 4-Hydroxyphenylcyclohexane
PubChem CID 14327
IUPAC Name 4-cyclohexylphenol
SMILES C1CCC(CC1)C2=CC=C(C=C2)O

Specifications

Melting Point 130°C
Color White
Quantity 25 g
Formula Weight 176.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyclohexylphenol
EINECSNumber (4)-0021
TSCA Yes