missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanophenylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

Catalog No. AAH58211AD
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH58211AD 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH58211AD Supplier Thermo Scientific Chemicals Supplier No. H58211AD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 131379-14-1 | C7H4BrNZn | 247.4 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyanophenylzinc bromide is used to produce biphenyl-4-carbonitrile.

Solubility
Reacts with water.

Notes
Incompatible with oxidizing agents, air, bases and heat. Air Sensitive. Store under dry iner

Chemical Identifiers

CAS 131379-14-1
Molecular Formula C7H4BrNZn
Molecular Weight (g/mol) 247.4
MDL Number MFCD00671984
InChI Key HEOMKTNNZDZMJZ-UHFFFAOYSA-M
Synonym zinc 2+ ion 4-cyanobenzen-1-ide bromide, zinc bromide 4-cyanobenzen-1-ide 1/1/1
PubChem CID 54750729
IUPAC Name zinc;benzonitrile;bromide
SMILES C1=CC(=CC=[C-]1)C#N.[Zn+2].[Br-]

Specifications

Density 0.98
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive; May form a precipitate
Solubility Information Reacts with water.
Formula Weight 247.4
Chemical Name or Material 4-Cyanophenylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Hazard Category H225-H260-H302-H314-H335-H336-H351
Hazard Statement GHS H Statement
H225-H261-H300-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Fatal if swallowed.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only