missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanophenylhydrazine hydrochloride, 97+%

Catalog No. AC392300250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC392300250 25 g Glass bottle
AC392300050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC392300250 Supplier Thermo Scientific Chemicals Supplier No. 392300250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2863-98-1 | C7H7N3 | 133.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2863-98-1
Molecular Formula C7H7N3
Molecular Weight (g/mol) 133.15
MDL Number MFCD00673994
InChI Key DZUUSHCOMPROCJ-UHFFFAOYSA-N
Synonym 4-cyanophenylhydrazine hydrochloride, 4-hydrazinylbenzonitrile hydrochloride, 4-hydrazinobenzonitrile hydrochloride, 4-cyanophenylhydrazine hcl, benzonitrile, 4-hydrazino-, monohydrochloride, 4-hydrazino-benzonitrile; hydrochloride, pubchem13684, 4-cyanophenylhydrazine-hcl, acmc-1co7p, ksc205o1j
PubChem CID 16212962
SMILES NNC1=CC=C(C=C1)C#N

Specifications

CAS Min % 97.0
CAS Max % 100.0
Melting Point 249.0°C
Color Brown to Orange
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (HPLC)
Packaging Glass bottle
Linear Formula NCC6H4NHNH2·HCl
Quantity 25 g
Solubility Information Solubility in water: soluble
IUPAC Name 4-hydrazinylbenzonitrile;hydrochloride
Formula Weight 169.61
Percent Purity 97+%
Physical Form Powder
Chemical Name or Material 4-Cyanophenylhydrazine hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Harmful in contact with skin.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fre

RUO – Research Use Only