missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanophenol, 99%

Catalog No. AC110831000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC110831000 100 g Glass bottle
AC110830250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC110831000 Supplier Thermo Scientific Chemicals Supplier No. 110831000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 767-00-0 | C7H5NO | 119.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 767-00-0
Molecular Formula C7H5NO
Molecular Weight (g/mol) 119.12
MDL Number MFCD00002312
InChI Key CVNOWLNNPYYEOH-UHFFFAOYSA-N
Synonym 4-cyanophenol, p-hydroxybenzonitrile, p-cyanophenol, benzonitrile, 4-hydroxy, benzonitrile, p-hydroxy, 4-cyano phenol, 4-hydroxybenzoic acid nitrile, 4-hydroxy-benzonitrile, 4-hydroxy benzonitrile, 4-hydroxybenzenecarbonitrile
PubChem CID 13019
ChEBI CHEBI:38622
IUPAC Name 4-hydroxybenzonitrile
SMILES C1=CC(=CC=C1C#N)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 110°C to 113°C
Color Beige-Gray to White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula NCC6H4OH
Quantity 100 g
Beilstein 10, 167
Solubility Information Solubility in water: slightly soluble
Formula Weight 119.12
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyanophenol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/
EINECSNumber 212-175-2

RUO – Research Use Only