missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanobiphenyl 98.0+%, TCI America™

Catalog No. C12995G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C12995G 5 g
C129925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C12995G Supplier TCI America Supplier No. C12995G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2920-38-9
Molecular Formula C13H9N
Molecular Weight (g/mol) 179.22
MDL Number MFCD00001821
InChI Key BPMBNLJJRKCCRT-UHFFFAOYSA-N
Synonym 4-cyanobiphenyl, 1,1'-biphenyl-4-carbonitrile, 4-biphenylcarbonitrile, p-phenylbenzonitrile, p-cyanobiphenyl, biphenyl-4-carbonitrile, 4-phenylbenzenecarbonitrile, 4-biphenylcarboxylic acid nitrile, 4-cyanodiphenyl
PubChem CID 18021
IUPAC Name [1,1'-biphenyl]-4-carbonitrile
SMILES N#CC1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 87°C
Color White-Yellow
Boiling Point 193°C
Quantity 5 g
UN Number 3439
Formula Weight 179.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyanobiphenyl