missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanobenzophenone, 98%

Catalog No. AAA1576414
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1576414 25 g
AAA1576403 1 g
AAA1576406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1576414 Supplier Thermo Scientific Chemicals Supplier No. A1576414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1503-49-7 | C14H9NO | 207.232 g/mol

Substituted benzophenones are used to determine the quantum yields for norbornadiene(N)quadricyclane(Q) and QN isomerization. It is employed as a triplet sensitizers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Substituted benzophenones are used to determine the quantum yields for norbornadiene(N)→quadricyclane(Q) and Q→N isomerization. It is employed as a triplet sensitizers.

Solubility
Insoluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.

Chemical Identifiers

CAS 1503-49-7
Molecular Formula C14H9NO
Molecular Weight (g/mol) 207.232
MDL Number MFCD00016382
InChI Key YSZWJJANSNFQMM-UHFFFAOYSA-N
PubChem CID 73921
IUPAC Name 4-benzoylbenzonitrile
SMILES C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)C#N

Specifications

Melting Point 110°C to 114°C
Quantity 25 g
UN Number UN3439
Beilstein 641181
Synonym 4-cyanobenzophenone, 4-benzoyl-benzonitrile, benzonitrile, 4-benzoyl, 4-cyano-benzophenon, 4-cyano-benzophenone, acmc-20amk8, #, benzonitrile,4-benzoyl, 4-phenylcarbonyl benzenecarbonitrile
Solubility Information Insoluble in water.
Formula Weight 207.23
Percent Purity 98%
Chemical Name or Material 4-Cyanobenzophenone
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 216-126-6
TSCA No

RUO – Research Use Only