missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanobenzoic acid, 98%

CAS: 619-65-8 | C8H5NO2 | 147.133 g/mol

Supplier:  Thermo Scientific Chemicals A1543414

 View more versions of this product

Catalog No. AAA1543414


Only null left
Explore available promotions

Description

Description

4-Cyanobenzoic acid is used as an inhibitor. 4-cyanobenzaldehyde and 4-cyanobenzoic acid can inhibit both the monophenolase activity and the diphenolase activity of mushroom tyrosinase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyanobenzoic acid is used as an inhibitor. 4-cyanobenzaldehyde and 4-cyanobenzoic acid can inhibit both the monophenolase activity and the diphenolase activity of mushroom tyrosinase.

Solubility
Soluble in methanol.

Notes
Air Sensitive. Incompatible with strong oxidizing agents, strong acids, strong bases.
Specifications

Specifications

4-Cyanobenzoic acid
∼221°C (decomposition)
MFCD00002528
1862865
p-cyanobenzoic acid, benzoic acid, 4-cyano, p-carboxybenzonitrile, 4-carboxybenzonitrile, benzoic acid, p-cyano, 4-cyano-benzoic acid, 4-cyano benzoic acid, benzoicacid, 4-cyano, 4-cyanobenzoicacid, para-cyanobenzoic acid
ADCUEPOHPCPMCE-UHFFFAOYSA-N
4-cyanobenzoic acid
12087
98%
619-65-8
C8H5NO2
25 g
Air Sensitive
Soluble in methanol.
C1=CC(=CC=C1C#N)C(=O)O
147.133
147.13

Chemical Identifiers

619-65-8
147.133
ADCUEPOHPCPMCE-UHFFFAOYSA-N
12087
C1=CC(=CC=C1C#N)C(=O)O
C8H5NO2
MFCD00002528
p-cyanobenzoic acid, benzoic acid, 4-cyano, p-carboxybenzonitrile, 4-carboxybenzonitrile, benzoic acid, p-cyano, 4-cyano-benzoic acid, 4-cyano benzoic acid, benzoicacid, 4-cyano, 4-cyanobenzoicacid, para-cyanobenzoic acid
4-cyanobenzoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

EINECSNumber : 210-606-9

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only