missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanobenzoic acid, 98%

Catalog No. p-7022586
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1543406 5 g
AAA1543414 25 g
AAA1543422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1543406 Supplier Thermo Scientific Chemicals Supplier No. A1543406
Only null left

CAS: 619-65-8 | C8H5NO2 | 147.133 g/mol

4-Cyanobenzoic acid is used as an inhibitor. 4-cyanobenzaldehyde and 4-cyanobenzoic acid can inhibit both the monophenolase activity and the diphenolase activity of mushroom tyrosinase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyanobenzoic acid is used as an inhibitor. 4-cyanobenzaldehyde and 4-cyanobenzoic acid can inhibit both the monophenolase activity and the diphenolase activity of mushroom tyrosinase.

Solubility
Soluble in methanol.

Notes
Air Sensitive. Incompatible with strong oxidizing agents, strong acids, strong bases.

Chemical Identifiers

CAS 619-65-8
Molecular Formula C8H5NO2
Molecular Weight (g/mol) 147.133
MDL Number MFCD00002528
InChI Key ADCUEPOHPCPMCE-UHFFFAOYSA-N
Synonym p-cyanobenzoic acid, benzoic acid, 4-cyano, p-carboxybenzonitrile, 4-carboxybenzonitrile, benzoic acid, p-cyano, 4-cyano-benzoic acid, 4-cyano benzoic acid, benzoicacid, 4-cyano, 4-cyanobenzoicacid, para-cyanobenzoic acid
PubChem CID 12087
IUPAC Name 4-cyanobenzoic acid
SMILES C1=CC(=CC=C1C#N)C(=O)O

Specifications

Melting Point ∼221°C (decomposition)
Quantity 5 g
Beilstein 1862865
Sensitivity Air Sensitive
Solubility Information Soluble in methanol.
Formula Weight 147.13
Percent Purity 98%
Chemical Name or Material 4-Cyanobenzoic acid
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 210-606-9
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only