accessibility menu, dialog, popup

UserName

HOON

CAS RN 619-65-8

IUPAC Name: 4-cyanobenzoic acid
Synonyms: P-cyanobenzoic acid
Molecular Weight (g/mol): 147.13
Molecular Formula: C8H5NO2
InChi Key: ADCUEPOHPCPMCE-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)C#N
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (4)