missing translation for 'onlineSavingsMsg'
Learn More

4'-Cyanoacetophenone 97.0+%, TCI America™

Catalog No. C114110G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C114110G 10 g
C114125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C114110G Supplier TCI America Supplier No. C114110G

Chemical Identifiers

CAS 1443-80-7
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.161
MDL Number MFCD00001825
InChI Key NLPHXWGWBKZSJC-UHFFFAOYSA-N
Synonym 4'-cyanoacetophenone, benzonitrile, 4-acetyl, p-cyanoacetophenone, 4-cyanoacetophenone, p-acetylbenzonitrile, 4-acetyl-benzonitrile, benzonitrile, p-acetyl, 4-acetylbenzenecarbonitrile, 4-acetylbenznitrile
PubChem CID 74044
IUPAC Name 4-acetylbenzonitrile
SMILES CC(=O)C1=CC=C(C=C1)C#N

Specifications

Melting Point 56°C
Color White-Yellow
Boiling Point 96°C
Quantity 10 g
UN Number 3439
Formula Weight 145.16
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Cyanoacetophenone