missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-3-iodopyridine, 98%, Thermo Scientific Chemicals

Catalog No. AAH2724703
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2724703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2724703 Supplier Thermo Scientific Chemicals Supplier No. H2724703

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10386-28-4 | Molecular Formula: C6H3IN2 | Molecular Weight: 230.008 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10386-28-4
Molecular Formula C6H3IN2
Molecular Weight (g/mol) 230.008
MDL Number MFCD08729285
InChI Key KMFIQUYSWTUCGD-UHFFFAOYSA-N
Synonym 3-iodoisonicotinonitrile, 4-cyano-3-iodopyridine, 3-iodo-4-pyridinecarbonitrile, 3-iodo-isonicotinonitrile, acmc-1c8uo, 4-pyridinecarbonitrile,3-iodo, 3-iodanylpyridine-4-carbonitrile
PubChem CID 12180887
IUPAC Name 3-iodopyridine-4-carbonitrile
SMILES C1=CN=CC(=C1C#N)I

Specifications

Melting Point 119°C to 122°C
Quantity 1 g
UN Number UN3439
Sensitivity Light sensitive
Formula Weight 230.01
Percent Purity 98%
Chemical Name or Material 4-Cyano-3-iodopyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only