missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-3-fluorophenyl 4-Butylbenzoate 98.0+%, TCI America™

Catalog No. C33411G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C33411G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C33411G Supplier TCI America Supplier No. C33411G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 86776-52-5
Melting Point 8°C
Color Yellow
Molecular Formula C18H16FNO2
Quantity 1 g
MDL Number MFCD11053362
Synonym 4-butylbenzoic acid 4-cyano-3-fluorophenyl ester, 4-butyl-benzoic acid 4-cyano-3-fluoro-phenyl ester, 4-cyano-3-fluorophenyl=4-butylbenzoate, 4-cyano-3-fluorophenyl 4-n-butylbenzoate, benzoic acid, 4-butyl-, 4-cyano-3-fluorophenyl ester, benzoic acid, 4-butyl-,4-cyano-3-fluorophenyl ester
InChI Key IBUUMDHMTARCSQ-UHFFFAOYSA-N
SMILES CCCCC1=CC=C(C=C1)C(=O)OC2=CC(=C(C=C2)C#N)F
IUPAC Name (4-cyano-3-fluorophenyl) 4-butylbenzoate
Molecular Weight (g/mol) 297.329
PubChem CID 13742342
Formula Weight 297.33
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Cyano-3-fluorophenyl 4-Butylbenzoate
Show More Show Less