missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-2'-nitrodiphenyl 98.0+%, TCI America™

Catalog No. C13001G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13001G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C13001G Supplier TCI America Supplier No. C13001G
Only null left

Chemical Identifiers

CAS 75898-34-9
Molecular Formula C13H8N2O2
Molecular Weight (g/mol) 224.219
MDL Number MFCD00191622
InChI Key LZUDBRIMDISCJM-UHFFFAOYSA-N
Synonym 4-Cyano-2′C-nitrobiphenyl, 2′C-Nitrobiphenyl-4-carbonitrile
PubChem CID 11851114
IUPAC Name 4-(2-nitrophenyl)benzonitrile
SMILES C1=CC=C(C(=C1)C2=CC=C(C=C2)C#N)[N+](=O)[O-]

Specifications

Melting Point 148°C
Color Yellow
Quantity 1 g
Formula Weight 224.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyano-2′-nitrodiphenyl