missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-2-fluorobenzoic Acid 97.0+%, TCI America™

Catalog No. C20511G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20511G 1 g
C20515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C20511G Supplier TCI America Supplier No. C20511G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 164149-28-4
Molecular Formula C8H4FNO2
Molecular Weight (g/mol) 165.12
MDL Number MFCD03094454
InChI Key KEJMSTJTAWACNI-UHFFFAOYSA-N
Synonym 2-fluoro-4-cyanobenzoic acid, benzoic acid, 4-cyano-2-fluoro, 4-cyano-2-fluoro-benzoic acid, 4-cyano-2-fluorobenzoicacid, pubchem4603, acmc-1bzbk, intermediates-zcf02232, 2-fluoro4-cyanobenzoic acid, ksc489o1d, 4-carboxy-3-fluorobenzonitrile
PubChem CID 590659
IUPAC Name 4-cyano-2-fluorobenzoic acid
SMILES OC(=O)C1=C(F)C=C(C=C1)C#N

Specifications

Melting Point 216°C
Color White
Quantity 1 g
UN Number 3439
Formula Weight 165.12
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyano-2-fluorobenzoic Acid