Learn More
4-Cyano-2-fluorobenzoic acid, 98%
CAS: 164149-28-4 | C8H4FNO2 | 165.12 g/mol
Supplier: Thermo Scientific Chemicals L1972214
Description
4-Cyano-2-fluorobenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Cyano-2-fluorobenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
Solubility
Insoluble in water.
Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
| 4-Cyano-2-fluorobenzoic acid | |
| 214°C to 218°C | |
| MFCD03094454 | |
| 8051929 | |
| Insoluble in water. | |
| OC(=O)C1=C(F)C=C(C=C1)C#N | |
| 165.12 | |
| 165.12 |
| 164149-28-4 | |
| C8H4FNO2 | |
| 25 g | |
| 2-fluoro-4-cyanobenzoic acid, benzoic acid, 4-cyano-2-fluoro, 4-cyano-2-fluoro-benzoic acid, 4-cyano-2-fluorobenzoicacid, pubchem4603, acmc-1bzbk, intermediates-zcf02232, 2-fluoro4-cyanobenzoic acid, ksc489o1d, 4-carboxy-3-fluorobenzonitrile | |
| KEJMSTJTAWACNI-UHFFFAOYSA-N | |
| 4-cyano-2-fluorobenzoic acid | |
| 590659 | |
| 98% |
Chemical Identifiers
| 164149-28-4 | |
| 165.12 | |
| KEJMSTJTAWACNI-UHFFFAOYSA-N | |
| 590659 | |
| OC(=O)C1=C(F)C=C(C=C1)C#N |
| C8H4FNO2 | |
| MFCD03094454 | |
| 2-fluoro-4-cyanobenzoic acid, benzoic acid, 4-cyano-2-fluoro, 4-cyano-2-fluoro-benzoic acid, 4-cyano-2-fluorobenzoicacid, pubchem4603, acmc-1bzbk, intermediates-zcf02232, 2-fluoro4-cyanobenzoic acid, ksc489o1d, 4-carboxy-3-fluorobenzonitrile | |
| 4-cyano-2-fluorobenzoic acid |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
H302-H315-H317-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only