missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-2-fluorobenzoic acid, 98%

Catalog No. p-7047761
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1972203 1 g
AAL1972206 5 g
AAL1972214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1972203 Supplier Thermo Scientific Chemicals Supplier No. L1972203
Only null left

CAS: 164149-28-4 | C8H4FNO2 | 165.12 g/mol

4-Cyano-2-fluorobenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyano-2-fluorobenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 164149-28-4
Molecular Formula C8H4FNO2
Molecular Weight (g/mol) 165.12
MDL Number MFCD03094454
InChI Key KEJMSTJTAWACNI-UHFFFAOYSA-N
Synonym 2-fluoro-4-cyanobenzoic acid, benzoic acid, 4-cyano-2-fluoro, 4-cyano-2-fluoro-benzoic acid, 4-cyano-2-fluorobenzoicacid, pubchem4603, acmc-1bzbk, intermediates-zcf02232, 2-fluoro4-cyanobenzoic acid, ksc489o1d, 4-carboxy-3-fluorobenzonitrile
PubChem CID 590659
IUPAC Name 4-cyano-2-fluorobenzoic acid
SMILES OC(=O)C1=C(F)C=C(C=C1)C#N

Specifications

Melting Point 214°C to 218°C
Quantity 1 g
Beilstein 8051929
Solubility Information Insoluble in water.
Formula Weight 165.12
Percent Purity 98%
Chemical Name or Material 4-Cyano-2-fluorobenzoic acid
Hazard Category H302-H315-H317-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only