accessibility menu, dialog, popup

UserName

NFOHO

CAS RN 164149-28-4

IUPAC Name: 4-cyano-2-fluorobenzoic acid
Synonyms: 4-cyano-2-fluorobenzoic acid
Molecular Weight (g/mol): 165.12
Molecular Formula: C8H4FNO2
InChi Key: KEJMSTJTAWACNI-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(F)C=C(C=C1)C#N
Percent Purity 
  • (2)
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (4)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)