missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-2-fluorobenzoic acid, 98%

CAS: 164149-28-4 | C8H4FNO2 | 165.12 g/mol

Supplier:  Thermo Scientific Chemicals L1972206

 View more versions of this product

Catalog No. AAL1972206


Only null left
Explore available promotions

Description

Description

4-Cyano-2-fluorobenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyano-2-fluorobenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

4-Cyano-2-fluorobenzoic acid
214°C to 218°C
MFCD03094454
8051929
Insoluble in water.
OC(=O)C1=C(F)C=C(C=C1)C#N
165.12
165.12
164149-28-4
C8H4FNO2
5 g
2-fluoro-4-cyanobenzoic acid, benzoic acid, 4-cyano-2-fluoro, 4-cyano-2-fluoro-benzoic acid, 4-cyano-2-fluorobenzoicacid, pubchem4603, acmc-1bzbk, intermediates-zcf02232, 2-fluoro4-cyanobenzoic acid, ksc489o1d, 4-carboxy-3-fluorobenzonitrile
KEJMSTJTAWACNI-UHFFFAOYSA-N
4-cyano-2-fluorobenzoic acid
590659
98%

Chemical Identifiers

164149-28-4
165.12
KEJMSTJTAWACNI-UHFFFAOYSA-N
590659
OC(=O)C1=C(F)C=C(C=C1)C#N
C8H4FNO2
MFCD03094454
2-fluoro-4-cyanobenzoic acid, benzoic acid, 4-cyano-2-fluoro, 4-cyano-2-fluoro-benzoic acid, 4-cyano-2-fluorobenzoicacid, pubchem4603, acmc-1bzbk, intermediates-zcf02232, 2-fluoro4-cyanobenzoic acid, ksc489o1d, 4-carboxy-3-fluorobenzonitrile
4-cyano-2-fluorobenzoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c

H302-H315-H317-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only