missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-2-fluorobenzoic acid, 97%, Thermo Scientific™

Numéro de catalogue. AC441830010
Modifier l'affichage
Click to view available options
Quantité:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
AC441830010 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. AC441830010 Fournisseur Thermo Scientific Chemicals Code fournisseur 441830010

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 164149-28-4
Formule moléculaire C8H4FNO2
Poids moléculaire (g/mol) 165.12
Numéro MDL MFCD03094454
Clé InChI KEJMSTJTAWACNI-UHFFFAOYSA-N
Synonyme 2-fluoro-4-cyanobenzoic acid, benzoic acid, 4-cyano-2-fluoro, 4-cyano-2-fluoro-benzoic acid, 4-cyano-2-fluorobenzoicacid, pubchem4603, acmc-1bzbk, intermediates-zcf02232, 2-fluoro4-cyanobenzoic acid, ksc489o1d, 4-carboxy-3-fluorobenzonitrile
PubChem CID 590659
Nom de l’IUPAC 4-cyano-2-fluorobenzoic acid
SOURIRES OC(=O)C1=C(F)C=C(C=C1)C#N

Spécifications

Nom ou substance chimique 4-Cyano-2-fluorobenzoic acid
CAS % minimum 96.0
CAS % maximal 100.0
Spectre infrarouge Authentic
Fourchette de pourcentages d’essai 96% min. (HPLC)
Emballage Glass bottle
Quantité 1g
Poids de formule 165.12
Pourcentage de pureté 97%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Contaminated work clothing should not be allowed out of the workplace.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only