missing translation for 'onlineSavingsMsg'
Learn More

4-(cis-4-Hydroxycyclohexyl)phenol 97.0+%, TCI America™

Catalog No. H13841G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13841G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H13841G Supplier TCI America Supplier No. H13841G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 370860-74-5
Molecular Formula C12H16O2
Molecular Weight (g/mol) 192.26
MDL Number MFCD03093649,MFCD23380209
InChI Key LSVLMFBVUQQWOQ-UHFFFAOYSA-N
PubChem CID 10176498
IUPAC Name 4-(4-hydroxycyclohexyl)phenol
SMILES OC1CCC(CC1)C1=CC=C(O)C=C1

Specifications

Melting Point 209°C
Color White
Quantity 1 g
Formula Weight 192.26
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(cis-4-Hydroxycyclohexyl)phenol