missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorothiobenzamide 97.0+%, TCI America™

Catalog No. C20535G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20535G 5 g
C205325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C20535G Supplier TCI America Supplier No. C20535G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2521-24-6
Molecular Formula C7H6ClNS
Molecular Weight (g/mol) 171.64
MDL Number MFCD00040956
InChI Key OKPUICCJRDBRJT-UHFFFAOYSA-N
Synonym 4-chlorothiobenzamide, 4-chlorobenzothioamide, p-chlorothiobenzamide, p-chlorobenzothiamide, 4-chlorobenzene-1-carbothioamide, 4-chloro-thiobenzamide, benzenecarbothioamide, 4-chloro, benzamide, p-chlorothio, benzamide, p-chlorothio-8ci, amino 4-chlorophenyl methane-1-thione
PubChem CID 2734826
IUPAC Name 4-chlorobenzene-1-carbothioamide
SMILES NC(=S)C1=CC=C(Cl)C=C1

Specifications

Melting Point 129°C
Quantity 5 g
Physical Form Crystal-Powder at 20°C
UN Number 2811
Formula Weight 171.64
Percent Purity ≥97.0% (HPLC,N)
Chemical Name or Material 4-Chlorothiobenzamide