missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorotetrahydro-2H-pyran 98.0+%, TCI America™

Catalog No. C22735G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22735G 5 g
C227325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C22735G Supplier TCI America Supplier No. C22735G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1768-64-5
Molecular Formula C5H9ClO
Molecular Weight (g/mol) 120.576
MDL Number MFCD00074911
InChI Key DHRSKOBIDIDMJZ-UHFFFAOYSA-N
Synonym 4-chlorotetrahydropyran, 4-chlorotetrahydro-2h-pyran, 2h-pyran, 4-chlorotetrahydro, 4-chloro-tetrahydro-pyran, 4-chloro-tetrahydro-2h-pyran, 4-chloranyloxane, 4-chlorotetrapyran, 4-chloro-tetrahydropyran, acmc-1bvar, bromomethylpentafluorobenzene
PubChem CID 137202
IUPAC Name 4-chlorooxane
SMILES C1COCCC1Cl

Specifications

Color Yellow
Boiling Point 150°C
Quantity 5 g
UN Number 1993
Formula Weight 120.58
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Chlorotetrahydro-2H-pyran