missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-4-Chlorostyrene oxide, 97+%

Catalog No. AAH2651614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2651614 25 g
AAH2651606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2651614 Supplier Thermo Scientific Chemicals Supplier No. H2651614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2788-86-5 | C8H7ClO | 154.593 g/mol

(±)-4-Chlorostyrene oxide is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+/-)-4-Chlorostyrene oxide is used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 2788-86-5
Molecular Formula C8H7ClO
Molecular Weight (g/mol) 154.593
MDL Number MFCD00956179
InChI Key IBWLXNDOMYKTAD-UHFFFAOYSA-N
Synonym 2-4-chlorophenyl oxirane, 4-chlorostyrene oxide, p-chlorostyrene oxide, oxirane, 4-chlorophenyl, p-chlorophenyl oxirane, 2-4-chloro-phenyl-oxirane, +/--4-chlorostyrene oxide, benzene, 1-chloro-4-epoxyethyl, 4-chlorophenyl oxirane, 4-chloro styrene oxide
PubChem CID 197010
IUPAC Name 2-(4-chlorophenyl)oxirane
SMILES C1C(O1)C2=CC=C(C=C2)Cl

Specifications

Density 1.283
Quantity 25 g
Refractive Index 1.552
Boiling Point 230°C
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 154.59
Percent Purity 98%
Chemical Name or Material (+/-)-4-Chlorostyrene oxide
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
RTECSNumber CZ0527000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only