missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloropyridine-2-carboxamide 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 162°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 156.57 |
| Percent Purity | ≥97.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Chloropyridine-2-carboxamide |
Safety and Handling
| TSCA | No |