missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. C14735G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C14735G Supplier TCI America Supplier No. C14735G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 4-Chlorophenylboronic Acid (contains varying amounts of Anhydride)
CAS 1679-18-1
Color White-Yellow
Quantity 5 g
Physical Form Crystal-Powder at 20°C
Molecular Formula C6H6BClO2
MDL Number MFCD00039137
Synonym 4-chlorophenyl boronic acid, 4-chlorobenzeneboronic acid, p-chlorophenylboronic acid, benzeneboronic acid, p-chloro, p-chlorobenzeneboronic acid, boronic acid, 4-chlorophenyl, boronic acid, p-chlorophenyl, 4-chlorophenyl boranediol, 4-chlorophenylbornic acid
InChI Key CAYQIZIAYYNFCS-UHFFFAOYSA-N
SMILES OB(O)C1=CC=C(Cl)C=C1
IUPAC Name (4-chlorophenyl)boronic acid
Molecular Weight (g/mol) 156.37
PubChem CID 74299
Formula Weight 156.37
Show More Show Less