missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenyl Methyl Sulfone 98.0+%, TCI America™

Catalog No. C12375G
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
C12375G 5 g
C123725G 25 g
2 options

Catalog No. C12375G

Supplier: TCI America C12375G

Only null left

Chemical Identifiers

CAS 98-57-7
Molecular Formula C7H7ClO2S
Molecular Weight (g/mol) 190.641
MDL Number MFCD00025067
InChI Key LMCOQDVJBWVNNI-UHFFFAOYSA-N
Synonym 4-chlorophenyl methyl sulfone, 1-chloro-4-methylsulfonyl benzene, p-chlorophenyl methyl sulfone, 4-methylsulfuryl chlorobenzene, benzene, 1-chloro-4-methylsulfonyl, methyl 4-chlorophenyl sulfone, sulfone, p-chlorophenyl methyl, 1-chloro-4-methylsulphonyl benzene, 4-chlorophenylmethyl sulfone, methyl p-chlorophenyl sulfone
PubChem CID 7395
IUPAC Name 1-chloro-4-methylsulfonylbenzene
SMILES CS(=O)(=O)C1=CC=C(C=C1)Cl

Specifications

Melting Point 98°C
Color White
Boiling Point 60°C
Quantity 5 g
Formula Weight 190.64
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chlorophenyl Methyl Sulfone
EINECSNumber (3)-3469
RTECSNumber WR5730000
TSCA Yes