missing translation for 'onlineSavingsMsg'
Learn More

4-Chloronitrobenzene 98.0+%, TCI America™

Catalog No. C0222500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0222500G 500 g
C022225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0222500G Supplier TCI America Supplier No. C0222500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 100-00-5
Molecular Formula C6H4ClNO2
Molecular Weight (g/mol) 157.553
MDL Number MFCD00007285
InChI Key CZGCEKJOLUNIFY-UHFFFAOYSA-N
Synonym 4-chloronitrobenzene, p-chloronitrobenzene, p-nitrochlorobenzene, benzene, 1-chloro-4-nitro, 4-nitrochlorobenzene, pncb, 4-chloro-1-nitrobenzene, p-nitrophenyl chloride, p-nitrochlorobenzol, p-nitroclorobenzene
PubChem CID 7474
ChEBI CHEBI:34399
IUPAC Name 1-chloro-4-nitrobenzene
SMILES C1=CC(=CC=C1[N+](=O)[O-])Cl

Specifications

Melting Point 84°C
Boiling Point 242°C
Quantity 500 g
UN Number 1578
Formula Weight 157.55
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloronitrobenzene
EINECSNumber (3)-0442
RTECSNumber CZ1050000
TSCA Yes