missing translation for 'onlineSavingsMsg'
Learn More

4-Chloromethyl-5-methyl-1,3-dioxol-2-one 94.0+%, TCI America™

Catalog No. C18825G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C18825G 5 g
C188225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C18825G Supplier TCI America Supplier No. C18825G

Chemical Identifiers

CAS 80841-78-7
Molecular Formula C5H5ClO3
Molecular Weight (g/mol) 148.542
MDL Number MFCD07787494
InChI Key QCLFSYYUWPUWQR-UHFFFAOYSA-N
Synonym 4-chloromethyl-5-methyl-1,3-dioxol-2-one, 4-cloromethyl-5-methyl-1,3-dioxol-2-one, 4-chloromethyl-5-methyl-1,3-dioxolen-2-one, 1,3-dioxol-2-one, 4-chloromethyl-5-methyl, 4-choromethyl-5-methyl-2-oxo-1,3-dioxole, 4-chloromethyl-5-methyl-2h-1,3-dioxol-2-one, pubchem10542, acmc-209pk7, ksc490e4r, 4-chloromethyl-5methyl-1,3-dioxol-2-one
PubChem CID 9855518
IUPAC Name 4-(chloromethyl)-5-methyl-1,3-dioxol-2-one
SMILES CC1=C(OC(=O)O1)CCl

Specifications

Color Yellow
Boiling Point 93°C
Quantity 5 g
Formula Weight 148.54
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Chloromethyl-5-methyl-1,3-dioxol-2-one
EINECSNumber (5)-6296
TSCA No