missing translation for 'onlineSavingsMsg'
Learn More

4-(Chloromethyl)-3,5-dimethylisoxazole 95.0+%, TCI America™

Catalog No. C22825G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22825G 5 g
C22821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C22825G Supplier TCI America Supplier No. C22825G
Only null left

Chemical Identifiers

CAS 19788-37-5
Molecular Formula C6H8ClNO
Molecular Weight (g/mol) 145.586
MDL Number MFCD00003154
InChI Key NIFAUKBQIAURIM-UHFFFAOYSA-N
Synonym 4-chloromethyl-3,5-dimethylisoxazole, isoxazole, 4-chloromethyl-3,5-dimethyl, 4-chloromethyl-3,5-dimethyl-1,2-oxazole, 4-chloromethyl-3,5-dimethyl-isoxazole, 3,5-dimethyl-4-chloromethylisoxazole, 4-chloromethyl-3,5-dimethyl isoxazole, pubchem8667, acmc-1c8qy, akos bc-3064
PubChem CID 88246
IUPAC Name 4-(chloromethyl)-3,5-dimethyl-1,2-oxazole
SMILES CC1=C(C(=NO1)C)CCl

Specifications

Boiling Point 88°C
Quantity 5 g
Formula Weight 145.59
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-(Chloromethyl)-3,5-dimethylisoxazole
TSCA Yes
Recommended Storage Refrigerator