missing translation for 'onlineSavingsMsg'
Learn More

4-(Chloromethyl)-2-(4-chlorophenyl)-1,3-thiazole, 97%, Thermo Scientific™

Numéro de catalogue. p-2700761
Modifier l'affichage
Click to view available options
Quantité:
1g
10g
Emballage:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CD03309DA 1g Amber Glass Bottle
CD03309EA 10g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CD03309DA Fournisseur Thermo Scientific Chemicals Code fournisseur CD03309DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 17969-22-1
Formule moléculaire C10H7Cl2NS
Poids moléculaire (g/mol) 244.133
Numéro MDL MFCD00047057
Clé InChI UEJQTBKTWJQBRN-UHFFFAOYSA-N
Synonyme 4-chloromethyl-2-4-chlorophenyl-1,3-thiazole, thiazole, 4-chloromethyl-2-4-chlorophenyl, 4-chloromethyl-2-4-chlorophenyl thiazole, thiazole,4-chloromethyl-2-4-chlorophenyl, 4-chloromethyl-2-4-chloro-phenyl-thiazole, thiazole, 4-chloromethyl-2-p-chlorophenyl, 4-chloromethyl-2-p-chlorophenylthiazole, 4-chloromethyl-2-4-chlorphenyl thiazole, 2-p-chlorophenyl-4-chloromethylthiazole
PubChem CID 610297
Nom de l’IUPAC 4-(chloromethyl)-2-(4-chlorophenyl)-1,3-thiazole
SOURIRES C1=CC(=CC=C1C2=NC(=CS2)CCl)Cl

Spécifications

Nom ou substance chimique 4-(Chloromethyl)-2-(4-chlorophenyl)-1, 3-thiazole
CAS % minimum 94.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 95%
Emballage Amber Glass Bottle
Quantité 1g
Poids de formule 244.14
Pourcentage de pureté 95%
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.