missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorocinnamic acid, predominantly trans, 99%

Catalog No. AAA1530422
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1530422 100 g
AAA1530414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1530422 Supplier Thermo Scientific Chemicals Supplier No. A1530422
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1615-02-7 | C9H7ClO2 | 182.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1615-02-7
Molecular Formula C9H7ClO2
Molecular Weight (g/mol) 182.60
MDL Number MFCD00004396
InChI Key GXLIFJYFGMHYDY-ZZXKWVIFSA-N
Synonym 4-chlorocinnamic acid, p-chlorocinnamic acid, 3-4-chlorophenyl acrylic acid, trans-4-chlorocinnamic acid, 2e-3-4-chlorophenyl prop-2-enoic acid, e-p-chlorocinnamic acid, cinnamic acid, p-chloro, trans-p-chlorocinnamic acid, 2-propenoic acid, 3-4-chlorophenyl, 3-p-chlorophenyl acrylic acid
PubChem CID 637797
ChEBI CHEBI:61116
IUPAC Name (E)-3-(4-chlorophenyl)prop-2-enoic acid
SMILES OC(=O)\C=C\C1=CC=C(Cl)C=C1

Specifications

Melting Point 248°C to 253°C
Quantity 100 g
Beilstein 1365308
Formula Weight 182.61
Percent Purity 99%
Chemical Name or Material 4-Chlorocinnamic acid, predominantly trans
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 216-564-8
RTECSNumber GD8490000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only