missing translation for 'onlineSavingsMsg'
Learn More

(4-Chlorobenzyl)triphenylphosphonium Chloride 98.0+%, TCI America™

Catalog No. C15815G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15815G 5 g
C158125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C15815G Supplier TCI America Supplier No. C15815G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1530-39-8
Molecular Formula C25H21Cl2P
Molecular Weight (g/mol) 423.32
MDL Number MFCD00041533
InChI Key RAHOAHBOOHXRDY-UHFFFAOYSA-M
Synonym 4-chlorobenzyl triphenylphosphonium chloride, 4-chlorobenzyl triphenylphosphoniumchloride, 4-chlorophenyl methyl triphenylphosphanium chloride, acmc-1bpq8, 4-chlorobenzyltriphenylphosphonium chloride, 4-chlorobenzyltriphenyl-phosphonium chloride, 4-chlorophenyl methyl-triphenylphosphanium chloride, 4-chlorophenyl methyl-triphenylphosphonium chloride, 4-chlorophenyl methyl triphenylphosphine, chloride, 4-chlorobenzyl triphenyl phosphonium chloride
PubChem CID 2733546
IUPAC Name [(4-chlorophenyl)methyl]triphenylphosphanium chloride
SMILES [Cl-].ClC1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1

Specifications

Color White
Quantity 5 g
Formula Weight 423.32
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (4-Chlorobenzyl)triphenylphosphonium Chloride