missing translation for 'onlineSavingsMsg'
Learn More

4-chlorure de chlorobenzoyle 98,0+%, TCI America™

Numéro de catalogue. C014125G
Modifier l'affichage
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
C014125G 25 g
C0141100G 100 g
C0141500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. C014125G Fournisseur TCI America Code fournisseur C014125G

Identifiants chimiques

CAS 122-01-0
Molecular Formula C7H4Cl2O
Molecular Weight (g/mol) 175.008
MDL Number MFCD00000686
InChI Key RKIDDEGICSMIJA-UHFFFAOYSA-N
Synonym benzoyl chloride, 4-chloro, p-chlorobenzoyl chloride, para-chlorobenzoyl chloride, 4-chlorobenzoic acid chloride, 4-chlorobenzoylchloride, 4-chloro benzoyl chloride, benzoylchloride, 4-chloro, benzoyl chloride, p-chloro, ccris 8935, pcboc
PubChem CID 8501
ChEBI CHEBI:60716
IUPAC Name 4-chlorobenzoyle chlorure
SMILES C1=CC(=CC=C1C(=O)Cl)Cl

Spécifications

Color Incolore
Boiling Point 222°C
Quantity 25 g
UN Number 3265
Formula Weight 175.01
Percent Purity ≥98.0% (GC,T)
Physical Form Liquide
Chemical Name or Material 4-Chlorobenzoyl Chloride
EINECSNumber (3)-0062&(3)-1409
RTECSNumber DM6635510
TSCA Yes