Learn More
4-Chlorobenzotrifluoride, 98+%
CAS: 98-56-6 | C7H4ClF3 | 180.554 g/mol
Supplier: Thermo Scientific Chemicals A151540E
Description
4-Chlorobenzotrifluoride acts as an important intermediate for dyes. It is used as an active component of polyurethane finishes. It is also used in adhesives, pigments, sealant chemicals, paints and coatings. Further, it is used as an environmental friendly solvent for cleaning or degreasing and dielectric fluid. It has a high capacity to dissolve inks, which is used in the printing industry. In addition to this, it is also used as xylene replacement in cleaners, thinners and other aromatic hydrocarbon blends.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Chlorobenzotrifluoride acts as an important intermediate for dyes. It is used as an active component of polyurethane finishes. It is also used in adhesives, pigments, sealant chemicals, paints and coatings. Further, it is used as an environmental friendly solvent for cleaning or degreasing and dielectric fluid. It has a high capacity to dissolve inks, which is used in the printing industry. In addition to this, it is also used as xylene replacement in cleaners, thinners and other aromatic hydrocarbon blends.
Solubility
Miscible with chloroform. Slightly miscible with water.
Notes
Store in a cool place. Light sensitive. Incompatible with sodium dimethyl sulfonate, strong bases and strong oxidizing agents.
Specifications
| 4-Chlorobenzotrifluoride | |
| -36°C | |
| 136°C to 138°C | |
| C7H4ClF3 | |
| MFCD00000627 | |
| UN2234 | |
| 14,2126 | |
| Miscible with chloroform. Slightly miscible with water. | |
| C1=CC(=CC=C1C(F)(F)F)Cl | |
| 180.554 | |
| 180.56 |
| 98-56-6 | |
| 1.346 | |
| 47°C (116°F) | |
| 1.446 | |
| 2500 g | |
| 510203 | |
| 4-chlorobenzotrifluoride, p-chlorobenzotrifluoride, 1-chloro-4-trifluoromethyl benzene, benzene, 1-chloro-4-trifluoromethyl, 4-chloro-alpha,alpha,alpha-trifluorotoluene, para-chlorobenzotrifluoride, p-chlorophenyl trifluoromethane, p-chloro-a,a,a-trifluorotoluene, pcbtf, p-chlorotrifluoromethylbenzene | |
| QULYNCCPRWKEMF-UHFFFAOYSA-N | |
| 1-chloro-4-(trifluoromethyl)benzene | |
| 7394 | |
| ≥98% |
Chemical Identifiers
| 98-56-6 | |
| 180.554 | |
| QULYNCCPRWKEMF-UHFFFAOYSA-N | |
| 7394 | |
| C1=CC(=CC=C1C(F)(F)F)Cl |
| C7H4ClF3 | |
| MFCD00000627 | |
| 4-chlorobenzotrifluoride, p-chlorobenzotrifluoride, 1-chloro-4-trifluoromethyl benzene, benzene, 1-chloro-4-trifluoromethyl, 4-chloro-alpha,alpha,alpha-trifluorotoluene, para-chlorobenzotrifluoride, p-chlorophenyl trifluoromethane, p-chloro-a,a,a-trifluorotoluene, pcbtf, p-chlorotrifluoromethylbenzene | |
| 1-chloro-4-(trifluoromethyl)benzene |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CHLOROBENZOTRIFLUORIDES
EINECSNumber : 202-681-1
RTECSNumber : XS9145000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only