accessibility menu, dialog, popup

UserName

FFFCl

CAS RN 98-56-6

IUPAC Name: 1-chloro-4-(trifluoromethyl)benzene
Synonyms: parachlorobenzotrifluoride
Molecular Weight (g/mol): 180.55
Molecular Formula: C7H4ClF3
InChi Key: QULYNCCPRWKEMF-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(Cl)C=C1
Boiling Point 
  • (2)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (4)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)