missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzotrifluoride, 98+%

Catalog No. p-7022396
Change view
Click to view available options
Quantity:
500 g
2500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1515436 500 g
AAA151540E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1515436 Supplier Thermo Scientific Chemicals Supplier No. A1515436
Only null left

CAS: 98-56-6 | C7H4ClF3 | 180.554 g/mol

4-Chlorobenzotrifluoride acts as an important intermediate for dyes. It is used as an active component of polyurethane finishes. It is also used in adhesives, pigments, sealant chemicals, paints and coatings. Further, it is used as an environmental friendly solvent for cleaning or degreasing and dielectric fluid. It has a high capacity to dissolve inks, which is used in the printing industry. In addition to this, it is also used as xylene replacement in cleaners, thinners and other aromatic hydrocarbon blends.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorobenzotrifluoride acts as an important intermediate for dyes. It is used as an active component of polyurethane finishes. It is also used in adhesives, pigments, sealant chemicals, paints and coatings. Further, it is used as an environmental friendly solvent for cleaning or degreasing and dielectric fluid. It has a high capacity to dissolve inks, which is used in the printing industry. In addition to this, it is also used as xylene replacement in cleaners, thinners and other aromatic hydrocarbon blends.

Solubility
Miscible with chloroform. Slightly miscible with water.

Notes
Store in a cool place. Light sensitive. Incompatible with sodium dimethyl sulfonate, strong bases and strong oxidizing agents.

Chemical Identifiers

CAS 98-56-6
Molecular Formula C7H4ClF3
Molecular Weight (g/mol) 180.554
MDL Number MFCD00000627
InChI Key QULYNCCPRWKEMF-UHFFFAOYSA-N
Synonym 4-chlorobenzotrifluoride, p-chlorobenzotrifluoride, 1-chloro-4-trifluoromethyl benzene, benzene, 1-chloro-4-trifluoromethyl, 4-chloro-alpha,alpha,alpha-trifluorotoluene, para-chlorobenzotrifluoride, p-chlorophenyl trifluoromethane, p-chloro-a,a,a-trifluorotoluene, pcbtf, p-chlorotrifluoromethylbenzene
PubChem CID 7394
IUPAC Name 1-chloro-4-(trifluoromethyl)benzene
SMILES C1=CC(=CC=C1C(F)(F)F)Cl

Specifications

Melting Point -36°C
Density 1.346
Boiling Point 136°C to 138°C
Flash Point 47°C (116°F)
Refractive Index 1.446
Quantity 500 g
UN Number UN2234
Beilstein 510203
Merck Index 14,2126
Solubility Information Miscible with chloroform. Slightly miscible with water.
Formula Weight 180.56
Percent Purity ≥98%
Chemical Name or Material 4-Chlorobenzotrifluoride
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CHLOROBENZOTRIFLUORIDES
EINECSNumber 202-681-1
RTECSNumber XS9145000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only