missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzhydrol 98.0+%, TCI America™

Catalog No. C0989500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0989500G 500 g
C098925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0989500G Supplier TCI America Supplier No. C0989500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 119-56-2
Molecular Formula C13H11ClO
Molecular Weight (g/mol) 218.68
MDL Number MFCD00004491
InChI Key AJYOOHCNOXWTKJ-UHFFFAOYSA-N
Synonym 4-chlorobenzhydrol, 4-chlorophenyl phenyl methanol, p-chlorobenzhydrol, chlorobenzhydrol, benzhydrol, p-chloro, 4-chlorodiphenylmethanol, benzhydrol, 4-chloro, 4-chlorophenyl phenylmethanol, 4-chlorobenzhydryl alcohol, benzenemethanol, 4-chloro-.alpha.-phenyl
PubChem CID 8401
ChEBI CHEBI:35091
IUPAC Name (4-chlorophenyl)-phenylmethanol
SMILES C1=CC=C(C=C1)C(C2=CC=C(C=C2)Cl)O

Specifications

Melting Point 61°C
Color White-Yellow
Boiling Point 151°C
Quantity 500 g
Formula Weight 218.68
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chlorobenzhydrol
EINECSNumber (9)-0364
TSCA Yes